Skip to Content
Merck
All Photos(1)

Key Documents

168017

Sigma-Aldrich

meso-2,3-Dimethylsuccinic acid

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$136.00

$136.00


Estimated to ship onApril 08, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$136.00

About This Item

Linear Formula:
HO2CCH(CH3)CH(CH3)CO2H
CAS Number:
Molecular Weight:
146.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$136.00


Estimated to ship onApril 08, 2025


Request a Bulk Order

Quality Level

Assay

99%

form

powder

mp

200 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

C[C@@H]([C@@H](C)C(O)=O)C(O)=O

InChI

1S/C6H10O4/c1-3(5(7)8)4(2)6(9)10/h3-4H,1-2H3,(H,7,8)(H,9,10)/t3-,4+

InChI key

KLZYRCVPDWTZLH-ZXZARUISSA-N

Application

meso-2,3-Dimethylsuccinic acid was used in the synthesis of:
  • meso-1,4-diiodo-2,3-dimethylbutane[1]
  • selenoacetal with an erythro-configured CHMe-CHMe fragment[2]
  • erythro-4-acetoxy-2,3-dimethylbutan-1-ol [2]
  • (±)-faranal[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of (3S*, 4R*, 6E, 10Z)-3, 4, 7, 11-tetramethyltrideca-6, 10-dienal (faranal) using stereospecific 1, 4-cis-hydrogenation of conjugated double bonds.
Vasil'ev AA, et al.
Journal of the Chemical Society. Perkin Transactions 1, 14, 2211-2216 (2000)
Highly selective formation of eight-membered-ring systems by oxidative cyclization with molybdenum pentachloride-an environmentally friendly and inexpensive access to 2,2'-cyclolignans.
Beate Kramer et al.
Angewandte Chemie (International ed. in English), 41(16), 2981-2982 (2002-08-31)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service