Skip to Content
Merck
All Photos(1)

Key Documents

D168009

Sigma-Aldrich

Dimethylmalonic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$56.90
25 G
$127.00

$56.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$56.90
25 G
$127.00

About This Item

Linear Formula:
(CH3)2C(COOH)2
CAS Number:
Molecular Weight:
132.11
Beilstein:
774375
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$56.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

crystals

mp

191-193 °C (lit.)

SMILES string

CC(C)(C(O)=O)C(O)=O

InChI

1S/C5H8O4/c1-5(2,3(6)7)4(8)9/h1-2H3,(H,6,7)(H,8,9)

InChI key

OREAFAJWWJHCOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabet Pires et al.
Physical chemistry chemical physics : PCCP, 22(42), 24351-24358 (2020-10-22)
The variation of the 31P chemical shift of triethylphosphine oxide in CDCl3 solution with a series of Brønsted acids at different molar ratios allows the determination of the value for the 1 : 1 species (δ1 : 1), which is much lower than the
P A Frey et al.
Science (New York, N.Y.), 264(5167), 1927-1930 (1994-06-24)
Spectroscopic properties of chymotrypsin and model compounds indicate that a low-barrier hydrogen bond participates in the mechanism of serine protease action. A low-barrier hydrogen bond between N delta 1 of His57 and the beta-carboxyl group of Asp102 in chymotrypsin can
M Phillips et al.
Clinical chemistry, 38(1), 60-65 (1992-01-01)
We describe a method for the collection and microanalysis of the volatile organic compounds in human breath. A transportable apparatus supplies subjects with purified air and samples their alveolar breath; the volatile organic compounds are captured in an adsorptive trap
J Schneede et al.
Analytical chemistry, 67(5), 812-819 (1995-03-01)
Methylmalonic acid (MMA) in serum is an established marker of cobalamin deficiency. MMA and other short-chain dicarboxylic acids react with 1-pyrenyldiazomethane to form stable, highly fluorescent 1-pyrenylmethyl monoesters. We have analyzed these esters in human blood by capillary electrophoresis (CE)
James R Harbridge et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 159(2), 195-206 (2002-12-17)
The effects of methyl rotation on electron spin-lattice relaxation times were examined by pulsed electron paramagnetic resonance for the major radicals in gamma-irradiated polycrystalline alpha-amino isobutyric acid, dimethyl-malonic acid, and L-valine. The dominant radical is the same in irradiated dimethyl-malonic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service