Skip to Content
Merck
All Photos(1)

Key Documents

D151408

Sigma-Aldrich

2,2-Dimethylbutane

99%

Synonym(s):

Neohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C(CH3)3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730736
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.97 (vs air)

Quality Level

vapor pressure

5.35 psi ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

797 °F

expl. lim.

~7.7 %

refractive index

n20/D 1.369 (lit.)

bp

50 °C (lit.)

mp

−100 °C (lit.)

density

0.649 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(C)C

InChI

1S/C6H14/c1-5-6(2,3)4/h5H2,1-4H3

InChI key

HNRMPXKDFBEGFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Some of the reported applications of 2,2-dimethylbutane include:
  • Palladium-catalyzed radical oxidative alkoxycarbonylation to prepare various alkyl carboxylates.
  • Iridium-iron-catalyzed tandem dehydrogenation-isomerization-hydrosilylation to synthesize corresponding silane.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Oxidative Alkane C? H Alkoxycarbonylation.
Lu L, et al.
Chemistry?A European Journal , 22(41), 14484-14488 (2016)
Conversion of alkanes to linear alkylsilanes using an iridium-iron-catalysed tandem dehydrogenation?isomerization?hydrosilylation.
Jia X and Huang Z
Nature Chemistry, 8(2), 157-161 (2016)
Yohan Lee et al.
Environmental science & technology, 49(14), 8899-8906 (2015-06-25)
The CH4/CO2 replacement that occurs in sH hydrates is investigated, with a primary focus on the enhanced CH4 recovery induced via structural transformation with a CO2 injection. In this study, neohexane (NH) is used as a liquid hydrocarbon guest in
Michael H Abraham et al.
European journal of medicinal chemistry, 43(3), 478-485 (2007-06-05)
Values of in vitro gas to lung partition coefficients, K(lung), of VOCs have been collected from the literature. For 44 VOCs, application of the Abraham solvation equation to log K(lung) yielded a correlation with R(2)=0.968 and S.D.=0.25 log units. Combination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service