Skip to Content
Merck
All Photos(1)

Key Documents

39760

Supelco

2,3-Dimethylbutane

analytical standard

Synonym(s):

Diisopropyl

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$93.50

$93.50


In StockDetails


Request a Bulk Order

Select a Size

Change View
5 ML
$93.50

About This Item

Linear Formula:
(CH3)2CHCH(CH3)2
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730737
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$93.50


In StockDetails


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

3 (vs air)

vapor pressure

4.54 psi ( 25.0 °C)(lit.)

Assay

≥99.5% (GC)

autoignition temp.

788 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

~7.7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.375 (lit.)
n20/D 1.375

bp

58 °C (lit.)

mp

−129 °C (lit.)

density

0.662 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CC(C)C(C)C

InChI

1S/C6H14/c1-5(2)6(3)4/h5-6H,1-4H3

InChI key

ZFFMLCVRJBZUDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Dimethylbutane is a chemical, which can undergo photo oxidation with nitrogen oxides to give carbon monoxide and acetone as the major products.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Montgomery.HJ
Groundwater Chemicals Desk Reference, Fourth Edition (2007)
2,3-Dimethylbutane. CAS# 79-29-8.
J B Galvin et al.
Journal of toxicology and environmental health. Part A, 58(1-2), 111-118 (1999-10-28)
M Imbriani
Giornale italiano di medicina del lavoro, 15(5-6), 133-138 (1993-09-01)
We have considered the opportunity of using some inert vapours (2-3 Dimethylbutane, 2 Methylpentane, n-Hexane) to measure the pulmonary capillary blood flow (Qc). The measurements were performed by means of a gas chromatograph connected with a mass spectrometer. The alpha
Xiaowu Chen et al.
Analytica chimica acta, 836, 61-71 (2014-07-01)
A novel method based on the strategy of N-phosphorylation labeling is described for quantification of twenty natural amino acids in human serum by reversed-phase liquid chromatography-electrospray tandem mass spectrometry (RP-LC/ESI-MS). The derivatization reaction was easily performed in one-pot reaction under

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service