Documents
CLS982522
Pyrex® culture tubes with screw-cap
O.D. × L 22 mm × 175 mm
Synonym(s):
Corning® 9825
About This Item
UNSPSC Code:
41121706
material
cap (Rubber Liner)
round bottom tube
packaging
case of 192 ea
pack of 48 ea
manufacturer/tradename
Corning 9825-22
O.D. × L
22 mm × 175 mm
General description
These Pyrex culture tubes are supplied with deep-form 18-415 GPI threaded phenolic caps to facilitate handling and sealing after autoclaving. Caps are fabricated from a special formula resistant to the effects of high temperatures and steam in autoclaving and are provided with a white rubber liner. The white enamel marking spot gives an excellent surface for pencil notations.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.