CLS808450
Pyrex® centrifuge tubes with pennyhead stopper
capacity 50 mL, With graduations
Synonym(s):
Corning® 8084
Select a Size
$1,020.00
Select a Size
About This Item
$1,020.00
Recommended Products
material
borosilicate glass
conical bottom tube
sterility
non-sterile
feature
cap: no
packaging
pack of 1
case of 12
manufacturer/tradename
Corning 8084-50
availability
not available in EU
O.D. × L
28 mm × 151 mm
capacity
50 mL
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service