CLS7995150
Pyrex® hybridization tubes, graduated with screw cap
I.D. × L 35 mm × 150 mm
Synonym(s):
Corning® 7995
Select a Size
$205.00
Select a Size
About This Item
$205.00
Recommended Products
material
borosilicate glass
cap (Plug seal)
flat bottom tube
sterility
non-sterile
feature
graduations
packaging
case of 2
pack of 2
manufacturer/tradename
Corning 7995-150
I.D. × L
35 mm × 150 mm
cap diam.
54 mm
cap thread
45
color
clear
capacity
100 mL
suitability
suitable for (for use in hybridization chambers)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service