CLS763016
Pyrex® PTFE key-hole stopper, color-coded
blue, stopper size 16
Synonym(s):
flask stopper, flask stoppers, stoppers, Corning® 7630
About This Item
Recommended Products
stopper size
16
manufacturer/tradename
Corning 7630-16
color
blue
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service