Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
PTFE key-hole stopper, color-coded
Synonym(s):
Corning
®
7630
Products
Product Category
stoppers (5)
Color
Manufacturer
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CLS763013
Pyrex
®
PTFE key-hole stopper, color-coded,
orange, stopper size 13
Expand
View Pricing
CLS76309
Pyrex
®
PTFE key-hole stopper, color-coded,
black, stopper size 9
Expand
View Pricing
CLS763019
Pyrex
®
PTFE key-hole stopper, color-coded,
green, stopper size 19
Expand
View Pricing
CLS763022
Pyrex
®
PTFE key-hole stopper, color-coded,
yellow, stopper size 22
Expand
View Pricing
CLS763016
Pyrex
®
PTFE key-hole stopper, color-coded,
blue, stopper size 16
Expand
View Pricing