Skip to Content
Merck
All Photos(1)

Key Documents

CLS6949

Corning® organic chemistry glassware kit with 24/40 joints

borosilicate glass (code 7740), non-sterile

Synonym(s):

Corning® 6949

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,630.00

$1,630.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$1,630.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

$1,630.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

borosilicate glass (code 7740)

sterility

non-sterile

manufacturer/tradename

Corning 6949

General description

This basic Corning® kit is supplied complete with drawer-size polyethylene case in which components are mounted in foam. The kit′s PYREX® components all use 24/40 Standard Taper joints for standardization.

The kit′s versatility enables many standard laboratory operations to be performed, such as: distillation, reaction with gas evolution, preparation or recovery, simple reflux, reflux with addition, fractional distillation and vacuum distillation. The case is approximately 130 mm x 256 mm x 343 mm.

Components

Components:
1.
Liebig condenser column
2. Cold-finger condenser
3. Round-bottom boiling flask, 100 mL
4. Round-bottom boiling flask, 250 mL
5. Round-bottom boiling flask, 500 mL
6. Graduated separatory funnel, 100 mL
7. PYREX® stopper
8. Reaction-vessel tube
9. Drying tube
10. Adapter outlet tube
11. Distilling tube adapter
12. Connecting tube adapters (2 each)

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service