CLS566050
Pyrex® Lifetime-Red™ volumetric flask with Pyrex® stopper
capacity 50 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass, Corning® 5660
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
borosilicate glass
long
Joint
joint stopper9
sterility
non-sterile
feature
closure type ST stopper
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 5660-50
tol. measuring range
0.05 mL
capacity
50 mL
Related Categories
Features and Benefits
- Pyrex® Lifetime-Red™ volumetric flasks with Pyrex® stoppers (volume 50 mL) have the capacity mark etched as a fine line through a narrow red band into clear glass, making a filler unnecessary and giving high legibility, accuracy and permanence.
- The strength of these flasks has been increased appreciably through machine-blown bodies to which are sealed heavy-tubing necks tooled for No. 9 Standard Taper glass stoppers.
- These Class A volumetric flasks have been manufactured to Class A tolerances as established by ASTM E- 694 for volumetric ware, ASTM E-542 for calibration of volumetric ware and ASTM E-288 for volumetric flasks. Calibrated.
Legal Information
Corning is a registered trademark of Corning, Inc.
Lifetime-Red is a trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)