CLS564410
Pyrex® volumetric flask with PTFE stopper
capacity 10 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass, Corning® 5644
About This Item
Recommended Products
material
borosilicate glass
long
stopper size
9
sterility
non-sterile
feature
class A
closure type Polytetrafluoroethylene stoppper
packaging
case of 12
manufacturer/tradename
Corning 5644-10
tol. measuring range
0.02 mL
capacity
10 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service