Key Documents
CLS5400250
Pyrex® iodine determination flask, with Pyrex® ST stopper
capacity 250 mL
Synonym(s):
flask erlenmeyer, flasks erlenmeyer, glass flasks, Corning® 5400
Select a Size
1 EA
$234.00
8 EA
$310.00
$234.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Select a Size
Change View
1 EA
$234.00
8 EA
$310.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$234.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
material
borosilicate glass
wide-mouth
Joint
joint ST/NS stopper22
sterility
non-sterile
feature
cap: no
closure type ST stopper
packaging
pack of 1
case of 8
manufacturer/tradename
Corning 5400-250
capacity
250 mL
General description
Flasks are blown in specillay designed molds, thereby insuring uniformity and proper contour. The stopper projects above the liquid seal trough to facilitate removal.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)