Key Documents
CLS4450125
Pyrex® baffled shaker flasks with heavy tooled rim
capacity 125 mL
Synonym(s):
Corning container
Select a Size
6 EA
$330.00
$330.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
6 EA
$330.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$330.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
narrow-mouth
sterility
non-sterile
feature
cap: no
closure type Rubber stopper
packaging
case of 6
manufacturer/tradename
Corning 4450-125
capacity
125 mL
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)