CLS3042100
Pyrex® Lifetime Red™ single metric scale graduated cylinder
volume 100 mL
Synonym(s):
Corning® 3042
Select a Size
$317.00
Select a Size
About This Item
$317.00
Recommended Products
feature
subdivision 1.0 mL
manufacturer/tradename
Corning 3042-100
tol. measuring range
0.6 mL
volume
100 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Calibrated "to contain"
- Single metric scale
- White enamel scale against red band
- Removable PE bumperguard
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service