CLS3022100
Pyrex® graduated cylinder, single metric scale, white enamel graduations
volume 100 mL
Synonym(s):
Corning® 3022
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
subdivision 1.0 mL
manufacturer/tradename
Corning 3022-100
tol. measuring range
0.6 mL
volume
100 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Calibrated "to contain"
- single metric scale
- Removable PE bumperguard
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service