Skip to Content
Merck
All Photos(2)

Key Documents

404764

Sigma-Aldrich

4-Acetamidobenzenesulfonyl azide

97%

Synonym(s):

p-ABSA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4SO2N3
CAS Number:
Molecular Weight:
240.24
Beilstein:
2219568
MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: click chemistry

mp

107-111 °C (lit.)

functional group

amide
azide

SMILES string

CC(=O)Nc1ccc(cc1)S(=O)(=O)N=[N+]=[N-]

InChI

1S/C8H8N4O3S/c1-6(13)10-7-2-4-8(5-3-7)16(14,15)12-11-9/h2-5H,1H3,(H,10,13)

InChI key

NTMHWRHEGDRTPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Acetamidobenzenesulfonyl azide is widely used as a reagent in organic synthesis to introduce azide groups into molecules.[1]

Application

Reagent for synthesis of:
Monosaccharide-derived alcohols
Non-peptidic NK3 receptor antagonists

Reagent for:
A late-stage intermolecular C-H olefination
Intramolecular isomuenchnone cycloaddition approach to antitumor agents
Rhodium-catalyzed carbene cyclization cycloaddition cascade reaction of vinylsulfonates
Suzuki-Miyaura cross coupling reaction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of novel aliphatic N-sulfonylamidino thymine derivatives by Cu (I)-catalyzed three-component coupling reaction
L Krstulovi, et al.
Croatica Chemica Acta. Arhiv Za Kemiju, 85, 525-534 (2012)
Organic Syntheses, 70, 93-93 (1992)
Synthetic Communications, 17, 1709-1709 (1987)
Tetrahedron, 49, 5109-5109 (1993)

Articles

Since the preparation of the first organic azide, phenyl azide, by Peter Griess in 1864 this energy-rich and versatile class of compounds has enjoyed considerable interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service