Skip to Content
Merck
All Photos(1)

Documents

CDS015197

Sigma-Aldrich

(R)-1-N-Boc-4-N-Fmoc-2-piperazine carboxylic acid

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28N2O6
Molecular Weight:
452.50
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

form

solid

functional group

Boc
Fmoc

SMILES string

CC(C)(C)OC(=O)N1CCN(C[C@@H]1C(O)=O)C(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C25H28N2O6/c1-25(2,3)33-24(31)27-13-12-26(14-21(27)22(28)29)23(30)32-15-20-18-10-6-4-8-16(18)17-9-5-7-11-19(17)20/h4-11,20-21H,12-15H2,1-3H3,(H,28,29)/t21-/m1/s1

InChI key

ZVHNNCSUTNWKFC-OAQYLSRUSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service