Skip to Content
Merck
All Photos(1)

Key Documents

676365

Sigma-Aldrich

4-Boc-piperazine-2-carboxylic acid

96%

Synonym(s):

1-Boc-piperazine-3-carboxylic acid, 4-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid, Piperazine-1,3-dicarboxylic acid 1-tert-butyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$95.10

$95.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$95.10

About This Item

Empirical Formula (Hill Notation):
C10H18N2O4
CAS Number:
Molecular Weight:
230.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$95.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

96%

form

powder or crystals

mp

231-239 °C

functional group

carboxylic acid

SMILES string

CC(C)(C)OC(=O)N1CCNC(C1)C(O)=O

InChI

1S/C10H18N2O4/c1-10(2,3)16-9(15)12-5-4-11-7(6-12)8(13)14/h7,11H,4-6H2,1-3H3,(H,13,14)

InChI key

YRYAXQJXMBETAT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service