Skip to Content
Merck
All Photos(2)

Key Documents

C68601

Sigma-Aldrich

3-Chloro-1-propanethiol

98%

Synonym(s):

γ-Chloropropyl mercaptan, 3-Chloropropan-1-thiol, 3-Chloropropanethiol, 3-Chloropropyl mercaptan

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$250.00

$250.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$250.00

About This Item

Linear Formula:
Cl(CH2)3SH
CAS Number:
Molecular Weight:
110.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$250.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.4921 (lit.)

bp

145.5 °C (lit.)

density

1.136 g/mL at 25 °C (lit.)

SMILES string

SCCCCl

InChI

1S/C3H7ClS/c4-2-1-3-5/h5H,1-3H2

InChI key

TZCFWOHAWRIQGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Chloro-1-propanethiol is a sulfur-containingcompound used as a building block in organic synthesis.It acts as a nucleophilic reagent in SN2 reactions. [1] [2].

Application

  • Solution-Phase Hybrid Passivation: 3-Chloro-1-propanethiolis used in the passivation of quantum dots to enhance the performance of infrared-band gap quantum dot solar cells, suggesting significant improvements in efficiency and stability (Mahajan et al., 2020).

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Staudinger reaction with 2-imino-1, 3-thiaselenanes toward the synthesis of C4 spiro-$\beta$-lactams
Toyoda, Yosuke and Ninomiya, et al.
Organic & Biomolecular Chemistry, 11, 2652-2659 (2013)
New ultrastable mesoporous adsorbent for the removal of mercury ions
De Canck, Els and Lapeire, et al.
Langmuir, 26, 10076-10083 (2010)
Arxel de León et al.
Journal of colloid and interface science, 456, 182-189 (2015-07-01)
The incorporation of gold nanoparticles in heterojunction solar cells is expected to increase the efficiency due to plasmon effects, but the literature studies are sometimes controversial. In this work, gold nanoparticles passivated with (Ph)n-(CH2)3SH (n=1, 2, 3) have been synthesized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service