Skip to Content
Merck
All Photos(1)

Key Documents

760714

Sigma-Aldrich

S-(4-Bromobutyl) thioacetate

96% (GC)

Synonym(s):

1-Butanethiol, 4-bromo-, acetate, Acetic acid, thio-,S-(4-bromobutyl) ester, Ethanethioic acid, S-(4-bromobutyl) ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$151.00

$151.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$151.00

About This Item

Empirical Formula (Hill Notation):
C6H11BrOS
CAS Number:
Molecular Weight:
211.12
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$151.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

96% (GC)

form

liquid

contains

copper as stabilizer

composition

(copper wire added to bulk material for stabilization)

storage condition

under inert gas

refractive index

n20/D 1.518

density

1.388 g/mL at 25 °C

storage temp.

−20°C

SMILES string

CC(=O)SCCCCBr

InChI

1S/C6H11BrOS/c1-6(8)9-5-3-2-4-7/h2-5H2,1H3

InChI key

ABARMQVSPZAYRI-UHFFFAOYSA-N

General description

S-(4-Bromobutyl) thioacetate is used as a precursor in the formation of organic self-assembled monolayers (SAMs) on metals. Deprotection reactions can then converts the thioacetate group to thiol. These are chemisorbed onto metal surfaces to form self-assembled monolayers with a high degree of structural order. They are often used as a molecular template to form functional nanostructures.[1]

Application

Used in the formation of Self-Assembled Monolayers on gold electrodes to modify its work function. [1][2]

Features and Benefits

High chemical stability in air or solution. Thioacetate prevents undesirable oxidation of thiol groups in oxygen containing atmosphere. Forms homogeneous interface structure in SAMs.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Thioacetate-Functionalized Self-Assembled Monolayers on Au: Toward High-Performance Ion-Selective Electrode for Ag
Jin, J., Zhou, W. J., Chen, Y., Liu, Y. L., Sun, X. Q., & Xi, H. T.
Bull. Korean Chem. Soc., 35(2), 601-601 (2014)
Formation and Structure of Self-Assembled Monolayers of Octylthioacetates on Au (111) in Catalytic Tetrabutylammonium Cyanide Solution
Park, T., Kang, H., Choi, I., Chung, H., Ito, E., Hara, M., & Noh, J.
Bull. Korean Chem. Soc., 30(2), 441-441 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service