Skip to Content
Merck
All Photos(2)

Key Documents

C64800

Sigma-Aldrich

4-Chlorophenyl glycidyl ether

99%

Synonym(s):

4-Chlorophenyl 2,3-epoxypropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9ClO2
CAS Number:
Molecular Weight:
184.62
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

solid

refractive index

n20/D 1.545 (lit.)

mp

31-33 °C (lit.)

SMILES string

Clc1ccc(OCC2CO2)cc1

InChI

1S/C9H9ClO2/c10-7-1-3-8(4-2-7)11-5-9-6-12-9/h1-4,9H,5-6H2

InChI key

KSLSZOOZWRMSAP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai Wu et al.
Applied microbiology and biotechnology, 99(22), 9511-9521 (2015-06-20)
Enantioselective hydrolysis of racemic epoxides mediated by epoxide hydrolases (EHs) is one of the most promising approaches to obtain enantiopure epoxides. In this study, we identified and characterized a novel EH (TpEH1) from Tsukamurella paurometabola by analyzing the conserved catalytic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service