Skip to Content
Merck
All Photos(1)

Key Documents

359556

Sigma-Aldrich

4-Azidoaniline hydrochloride

97%

Synonym(s):

4-Aminophenyl azide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$135.00
1 G
$400.00

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$135.00
1 G
$400.00

About This Item

Linear Formula:
N3C6H4NH2 · HCl
CAS Number:
Molecular Weight:
170.60
Beilstein:
7942921
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

reaction suitability

reaction type: click chemistry

mp

165 °C (dec.) (lit.)

functional group

azide

SMILES string

Cl.Nc1ccc(cc1)N=[N+]=[N-]

InChI

1S/C6H6N4.ClH/c7-5-1-3-6(4-2-5)9-10-8;/h1-4H,7H2;1H

InChI key

SDYAJRBHPPWHSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Azidoaniline hydrochloride is suitable for use in the preparation of photo-reactive polymers, poly(acrylic acid) having encorporated phenylazido groups.[1] It may be used in the preparation of 1-(4-aminophenyl)-1H-[1,2,3]-triazol-4-ylcarbonyl-Phe-Gly-Phe-Gly-OH.[2] It is suitable for use in the synthesis of photo-reactive polysaccharides.[3]

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Barbucci et al.
Biomaterials, 24(6), 915-926 (2002-12-31)
Surface microfabrication techniques were widely utilised for the spatial control of in vitro cell behaviour. A photo-immobilisation procedure was utilised to create micropatterned surfaces: four different stripe patterns (100, 50, 25 and 10 microm) of hyaluronan (Hyal) and its sulphated
D R Nisbet et al.
Journal of biomedical materials research. Part A, 89(1), 24-35 (2008-04-12)
In this study, thermoresponsive xyloglucan hydrogel scaffolds were investigated as candidates for neural tissue engineering of the spinal cord. The hydrogels were optimized to provide similar mechanical properties to that of native spinal cord, although also being functionalized through the
D Stangl et al.
Biochemistry, 30(35), 8605-8611 (1991-09-03)
Calcitonin gene-related peptide (CGRP) receptors were solubilized from human (h) cerebellum with use of the zwitterionic detergent 3-[(3-cholamidopropyl)dimethylammonio]-1-propanesulfonic acid (CHAPS). Scatchard analysis of equilibrium binding data indicated that the soluble extract contained a single class of CGRP binding sites with
Christian W Tornøe et al.
The Journal of organic chemistry, 67(9), 3057-3064 (2002-04-27)
The cycloaddition of azides to alkynes is one of the most important synthetic routes to 1H-[1,2,3]-triazoles. Here a novel regiospecific copper(I)-catalyzed 1,3-dipolar cycloaddition of terminal alkynes to azides on solid-phase is reported. Primary, secondary, and tertiary alkyl azides, aryl azides
Yoshihiro Ito et al.
Biomaterials, 24(18), 3021-3026 (2003-08-05)
A protein micro-array, called a "cell chip" was constructed by using a photo-reactive polymer for a cell-adhesion assay. Various amounts of albumin or fibronectin were covalently immobilized on a polystyrene dish using a micro-spotter with a dip pen. First, poly(acrylic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service