Skip to Content
Merck
All Photos(1)

Key Documents

H28203

Sigma-Aldrich

4-(2-Hydroxyethyl)morpholine

ReagentPlus®, 99%

Synonym(s):

2-Morpholinoethanol, 4-Morpholineethanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$57.60
100 G
$64.50
250 G
$75.70

$57.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$57.60
100 G
$64.50
250 G
$75.70

About This Item

Empirical Formula (Hill Notation):
C6H13NO2
CAS Number:
Molecular Weight:
131.17
Beilstein:
104375
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$57.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

product line

ReagentPlus®

Assay

99%

form

liquid

refractive index

n20/D 1.476 (lit.)

bp

227 °C/757 mmHg (lit.)

density

1.083 g/mL at 25 °C (lit.)

SMILES string

OCCN1CCOCC1

InChI

1S/C6H13NO2/c8-4-1-7-2-5-9-6-3-7/h8H,1-6H2

InChI key

KKFDCBRMNNSAAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter A Burland et al.
Bioorganic & medicinal chemistry, 19(18), 5679-5692 (2011-08-25)
In this work libraries of morpholines and oxazepanes have been prepared via the reductive amination reaction between dialdehydes, derived from carbohydrates, and a range of amines. In this way, functionalised morpholines and oxazepanes have been prepared that include N-alkylated derivatives
Antoine Frère et al.
Biomacromolecules, 16(3), 769-779 (2015-01-21)
RNAi therapeutics are promising therapeutic tools that have sparked the interest of many researchers. In an effort to provide a safe alternative to PEI, we have designed a series of new guanidinium- and morpholino-functionalized biocompatible and biodegradable polycarbonate vectors. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service