Skip to Content
Merck
All Photos(1)

Key Documents

C29002

Sigma-Aldrich

3-Chloro-1-butene

contains sodium/potassium carbonate as stabilizer, 97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$49.40
100 ML
$502.00

$49.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$49.40
100 ML
$502.00

About This Item

Linear Formula:
CH3CHClCH=CH2
CAS Number:
Molecular Weight:
90.55
Beilstein:
1679232
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$49.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

contains

sodium/potassium carbonate as stabilizer

refractive index

n20/D 1.419 (lit.)

bp

62-65 °C (lit.)

density

0.9 g/mL at 25 °C (lit.)

SMILES string

CC(Cl)C=C

InChI

1S/C4H7Cl/c1-3-4(2)5/h3-4H,1H2,2H3

InChI key

VZGLVCFVUREVDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-16.6 °F - closed cup

Flash Point(C)

-27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service