Skip to Content
Merck
All Photos(1)

Key Documents

28115

Sigma-Aldrich

Crotyl chloride

technical, (cis+trans), mixture of cis- and trans-isomers (~1:6), ~70% (GC)

Synonym(s):

cis,trans-1-Chloro-2-butene

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$362.00

$362.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$362.00

About This Item

Linear Formula:
CH3CH=CHCH2Cl
CAS Number:
Molecular Weight:
90.55
Beilstein:
605304
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$362.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical

Assay

~70% (GC)

form

liquid

impurities

~30% 3-chloro-1-butene

refractive index

n20/D 1.430

bp

85-86 °C (lit.)

solubility

water: soluble 14 g/L at 20 °C

density

0.923 g/mL at 20 °C (lit.)

functional group

alkyl halide
chloro

SMILES string

C\C=C\CCl

InChI

1S/C4H7Cl/c1-2-3-4-5/h2-3H,4H2,1H3/b3-2+

InChI key

YTKRILODNOEEPX-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Crotyl chloride reacts with trichlorosilane in the presence of tertiary amine and copper salts to give the corresponding allyltrichlorosilanes[1].

Application

Crotyl chloride has been used:
  • to investihgate the kinetics of the reactions of atomic chlorine with series of unsaturated aldehydes and ketones at 298K, to probe structure-reactivity relationships[2]
  • in the preparation of (E)-crotyltrichlorosilane[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereohomogeneous synthesis of (E)-and (Z)-crotyltrifluorosilanes and highly stereoselective allylation of aldehydes.
Kira M, et al.
Tetrahedron Letters, 30(9), 1099-1102 (1989)
Rate constants for the reactions of chlorine atoms with a series of unsaturated aldehydes and ketones at 298 K: structure and reactivity.
Wang W, et al.
Physical Chemistry Chemical Physics, 4(10), 1824-1831 (2002)
The condensation reaction of trichlorosilane with allylic chlorides catalyzed by copper salts in the presence of a tertiary amine.
Furuya N and Sukawa T.
Journal of Organometallic Chemistry, 96(1), C1-C3 (1975)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service