Skip to Content
Merck
All Photos(1)

Documents

C111201

Sigma-Aldrich

Cyclopentadecanone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H28(=O)
CAS Number:
Molecular Weight:
224.38
Beilstein:
1618444
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

bp

120 °C/0.3 mmHg (lit.)

mp

63-65 °C (lit.)

density

0.897 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCCCCCCCCCCC1

InChI

1S/C15H28O/c16-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-15/h1-14H2

InChI key

OSOIQJGOYGSIMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosaad A Abdel-Wahhab et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 141, 111399-111399 (2020-05-19)
This study aimed to determine the bioactive compounds of Bacillus sp. MERNA97 extract and to evaluate their efficacy against the oxidative damage, genotoxicity, chromosomal aberration and DNA fragmentation in rats treated with AFB1. Sixty male Sprague-Dawley rats were divided into
Simon Gane et al.
PloS one, 8(1), e55780-e55780 (2013-02-02)
Whether olfaction recognizes odorants by their shape, their molecular vibrations, or both remains an open and controversial question. A convenient way to address it is to test for odor character differences between deuterated and undeuterated odorant isotopomers, since these have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service