Merck
Molecular vibration-sensing component in human olfaction.
PloS one (2013-02-02)
Simon Gane, Dimitris Georganakis, Klio Maniati, Manolis Vamvakias, Nikitas Ragoussis, Efthimios M C Skoulakis, Luca Turin
ABSTRACT
Whether olfaction recognizes odorants by their shape, their molecular vibrations, or both remains an open and controversial question. A convenient way to address it is to test for odor character differences between deuterated and undeuterated odorant isotopomers, since these have identical ground-state conformations but different vibrational modes. In a previous paper (Franco et al. (2011) Proc Natl Acad Sci USA 108:9, 3797-802) we showed that fruit flies can recognize the presence of deuterium in odorants by a vibrational mechanism. Here we address the question of whether humans too can distinguish deuterated and undeuterated odorants. A previous report (Keller and Vosshall (2004) Nat Neurosci 7:4, 337-8) indicated that naive subjects are incapable of distinguishing acetophenone and d-8 acetophenone. Here we confirm and extend those results to trained subjects and gas-chromatography [GC]-pure odorants. However, we also show that subjects easily distinguish deuterated and undeuterated musk odorants purified to GC-pure standard. These results are consistent with a vibrational component in human olfaction.
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Cyclopentadecanone, 98%
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Acetophenone, ≥98%, FG