Key Documents
BR782881
BRAND® BRANDplates® for inserts
6-well with lid cellGrade S
Synonym(s):
BRANDplates®, Cell culture, Insert System, Inserts, Plates, Tissue culture
Select a Size
10 EA
$231.00
$231.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
10 EA
$231.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material:
polystyrene
transparent wells
size:
6 well
$231.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polystyrene
transparent wells
sterility
sterile
feature
lid
single packed
packaging
pack of 10 ea
pkg of Individually wrapped, with lid
manufacturer/tradename
BRAND 782881
size
6 well
suitability
suitable for (bacteriological assays)
General description
PS. pureGrade S surface, untreated, sterile, cover with condensation rings.Manufactured according to the requirements in ANSI/SLAS Standards 1 and 4.BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic
Features and Benefits
  • Non-treated surface
  • Side well access for easier pipetting and removing cover slips
  • Perfect positioning of the BRANDplates® insert
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)