Skip to Content
Merck
All Photos(1)

Key Documents

BR782035

BRAND® microplate BRANDplates®, cellGrade Plus

size 96 wells, size 330 μL, flat bottom, black, with transparent bottom

Synonym(s):

BRANDplates®, Cell culture, Microplates, Tissue culture, cellGradeplus

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$128.00

$128.00


Estimated to ship onApril 16, 2025



Select a Size

Change View
5 EA
$128.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
black wells
flat bottom
polystyrene
size:
330 μL
96 wells
working volume:
330 μL
sterility:
sterile
feature:
lid
skirt (F-bottom)

$128.00


Estimated to ship onApril 16, 2025


material

black wells
flat bottom
polystyrene

sterility

sterile

feature

lid
skirt (F-bottom)

packaging

pack of 5 ea
pkg of (bulk packed)

manufacturer/tradename

BRAND 782035

size

330 μL
96 wells

working volume

330 μL

color

black, with transparent bottom

Looking for similar products? Visit Product Comparison Guide

General description

For reduced-serum media cultivation of cells.For cultivation of fastidious cell lines.In addition to carboxyl and hydroxyl chemical groups, free amino groups are present on the surface.The surface has a protein-like composition, so cells can directly attach and spread out.Cells adhere faster, better rate of yield.Sensitive cell lines can be cultivated.Suited for serum reduced cultivation of cells.Clearly distinguishable through the color code: orange, embossed alphanumeric coding for 96-well standard plates.Free from endotoxins, DNase, DNA, RNase, non-cytotoxic.PS, sterile, with lids.BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic

Features and Benefits

• Optimal surfaces for optimal results• Reliable quality from the cleanroom• Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service