Skip to Content
Merck
All Photos(1)

Key Documents

BR781965

BRAND® microplate BRANDplates®, cellGrade

size 96 wells, size 350 μL, flat bottom, white

Synonym(s):

BRANDplates®, Cell culture, Microplates, Tissue culture, cellGrade

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
$507.00

$507.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
50 EA
$507.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
flat bottom
polystyrene
white wells
size:
350 μL
96 wells
sterility:
sterile
binding type:
cell culture treated
feature:
lid
skirt (F-bottom)

$507.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

flat bottom
polystyrene
white wells

sterility

sterile

feature

lid
skirt (F-bottom)

packaging

pack of 50 ea
pkg of (individually packed)

manufacturer/tradename

BRAND 781965

size

350 μL
96 wells

working volume

350 μL

color

white

binding type

cell culture treated

Looking for similar products? Visit Product Comparison Guide

General description

Standard plate for the cultivation of adherent cell lines.PS-surface with different chemical groups, such as carboxyl and hydroxyl groups, that are freely accessible.Surface is hydrophilic compared with non-treated PS.Serum components are easily bound onto the freely accessible chemical groups, allowing an indirect adhesion of cells.Clearly distinguishable through the color code: orange, embossed alphanumeric coding for 96-well standard plates.Free from endotoxins, DNase, DNA, RNase, non-cytotoxic.PS, sterile, with lids.BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic

Application

  • standard plate for the cultivation of adherent cells without additional coatings
  • cultivation of suspension cells
  • high content screenings
  • fluorescence assays
  • luminescence assays

Features and Benefits

  • BRAND® microplate BRANDplates®, cellGrade (96 Brand) offer precise measurement of samples with high accuracy.
  • Optimal surfaces for optimal results: High purity, crystal-clear polystyrene (PS)-surface with different chemical groups, such as carboxyl and hydroxyl groups, that are freely accessible.
  • Surface is hydrophilic compared with non-treated PS.
  • Reliable quality from the cleanroom: Automated, ISO class 7 cleanroom production ensures the best possible cleanliness.
  • Tested by independent laboratories.
  • The raw materials used in the plates fulfill the relevant requirements of the USP and ISO 10 993.
  • Serum components are easily bound onto the freely accessible chemical groups, allowing an indirect adhesion of cells.
  • Clearly distinguishable through the color code: orange, embossed alphanumeric coding for 96-well standard plates.
  • Free from endotoxins, DNase, DNA, RNase, non-cytotoxic.
  • PS, sterile, with lids.
  • Compatible with all ANSI/SLAS conforming analytic equipment.

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service