Documents
BR749311
BRAND® micro haematocrit capillary
sodium hep. w/o cal. mark I.D 1
Synonym(s):
Haematocrit capillaries
About This Item
UNSPSC Code:
41121500
NACRES:
NB.22
material
red
feature
DIN ISO 12772 and BS 4316-68, To Contain
packaging
pack of 1000 ea
manufacturer/tradename
BRAND 749311
General description
Compliant with DIN ISO 12772 and BS 4316-68 standards, these tubes feature a color code for easy identification and use in hematocrit centrifuges. The heparinized tubes are treated with sodium heparinate on the internal surface and are marked with a red color code, making them suitable for capillary blood samples. In contrast, the non-heparinized tubes are designated with a blue color code, specifically designed for use with heparinized venous blood.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Visit the Document Library
Customers Also Viewed
Slide 1 of 1
1 of 1
Patrick Siyambulela Mafunda et al.
Animal reproduction science, 224, 106664-106664 (2020-12-05)
Several standard descriptions of the avian male and female reproductive tract have been reported, including effects of age, stage of reproductive maturity and gonadal hormone concentrations. Limited information on penguin reproductive biology and a lack of information on the African