Skip to Content
Merck
All Photos(1)

Key Documents

BR707529

BRAND® titrating tube with screw cap for Titrette® 25mL / 50mL

with integrated discharge/recircirculation valve, from serial-number 01K

Synonym(s):

Accessories, Bottle-top Burette, Burettes, Titrette®

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$168.00

$168.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
1 EA
$168.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

$168.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


packaging

pack of 1 ea

manufacturer/tradename

BRAND 707529

General description

The BRAND® titrating tube with screw cap is specifically designed for use with the Titrette® 25 and 50 mL titrators. This tube provides a secure and reliable solution for conducting titrations, ensuring accurate measurements and ease of use. The screw cap ensures a tight seal, preventing leaks and contamination during the titration process.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Titrette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service