Key Documents
BR462617
BRAND® rubber conical gasket for filter funnels and filter flasks
size 28 mm
Synonym(s):
Filter, Rubber gasket
Select a Size
10 EA
$106.00
$106.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
10 EA
$106.00
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
$106.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
EPDM rubber
packaging
pack of 10 ea
manufacturer/tradename
BRAND 462617
size
28 mm
General description
Conical gaskets to place between filter funnel and filter flask.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)