Key Documents
BR36981
BRAND® volumetric flask BLAUBRAND® USP conformity certified
capacity 250 mL, neck joint: ST/NS 14/23, glass stopper
Synonym(s):
BLAUBRAND®, USP, Volumetric flasks
Select a Size
2 EA
$129.00
$129.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
2 EA
$129.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$129.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
glass stopper
Joint
joint: ST/NS neck14/23
feature
accuracy class A
packaging
pack of 2
manufacturer/tradename
BRAND 36981
capacity
250 mL
General description
The BRAND® volumetric flask BLAUBRAND® USP conformity certified is made from high-quality Boro 3.3 glass and complies with DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring accurate liquid measurement for a variety of laboratory applications. Each flask comes with a USP batch certificate, confirming its adherence to stringent quality and safety standards. This volumetric flask is an essential tool for precise analytical work, making it ideal for laboratories that require reliable and certified liquid handling solutions.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)