BR129748
BRAND® PE-LD Media Bottle with screw cap
capacity 250 mL
Select a Size
$15.30
Select a Size
About This Item
$15.30
Recommended Products
material
low-density polyethylene bottle
round bottle
Joint
Joints threaded neck (GL40)
sterility
non-sterile
feature
graduations: no
with cap
packaging
pack of 1 ea
manufacturer/tradename
BRAND 129748
capacity
250 mL
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)