BR129738
BRAND® PE-LD Media Bottle with screw cap
capacity 100 mL
Select a Size
$15.80
Estimated to ship onMarch 21, 2025
Select a Size
About This Item
$15.80
Estimated to ship onMarch 21, 2025
Recommended Products
material
low-density polyethylene bottle
round bottle
Joint
Joints threaded neck (GL32)
feature
graduations: no
with cap
packaging
pack of 1 ea
manufacturer/tradename
BRAND 129738
capacity
100 mL
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)