Documents
BAH420580001
SP Bel-Art® Dry-Keeper Horizontal Desiccator Cabinet
PVC, volume 2 ft3
About This Item
UNSPSC Code:
41123002
NACRES:
NB.43
material
PVC
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art H42058-0001
volume
2 ft3
General description
Clear and durable PVC desiccator cabinet has sleek black ABS accents, horizontal profile, and movable shelves for adjusting dry storage space configuration for moisture-sensitive materials.
  • Spacious 2 cu. ft. interior volume
  • Two adjustable, removable, and perforated shelves facilitate full circulation of air; shelf rails can be repositioned on any one of 8 steps set at 28mm intervals; additional shelves and rails sold separately
  • Large, in-door digital hygrometer for easy visualization of rH%
  • Gasketed door opens down and closes with two latches for a tight seal; interior opening measures 45 x 23cm (17¾ x 9")
  • Includes desiccant tray
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP