Skip to Content
Merck
All Photos(1)

Key Documents

BAF420741117

SP Bel-Art® Secador® 4.0 Vertical Auto-Desiccator Cabinet

internal volume 1.9 ft3, AC/DC input 120 V, US 3-pin plug, blue (with clear door)

Synonym(s):

Bel-Art® Secador® Auto Desiccator Cabinets: Vertical Models, SP BEL-ART SECADOR VERTICAL PROFILE BLUE 4.0 AUTO-DESICCATOR CABINET WITH CLEAR DOOR; 120V, 1.9 CU. FT.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104510

material

blue (with clear door)
polyester (Co-polyester)

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F42074-1117

internal volume

1.9 ft3

Looking for similar products? Visit Product Comparison Guide

General description

  • Polycarbonate construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Amber color models reduce visible light penetration by over 50%; perfect for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Automatic dehumidification and desiccant regeneration cycle operates continuously every 20 minutes
  • Color-indicating silica gel inside the desiccant regeneration module allows quick visual confirmation of desiccant status
  • Large door with easy-to-view digital hygrometer maximizes access to interior space; includes patented seal design, tab latches and security loops for a padlock or tamper-evident seals; opens right to left
  • Perforated shelves facilitate circulation of dry air
  • Choose from an extensive selection of sizes, styles and colors in 100, 120 or 230V (CE marked) models for use worldwide
  • 230V units include power cord/transformer that includes Q-NA, Q-SAA, Q-UK and Q-EU input connectors to accommodate North American, Australian, UK and European configurations
  • Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
  • Replacement Secador® Auto Desiccator Module Desiccant Beads, BAF420740020

Protecting valuable humidity-sensitive items, such as reagents or electronics, is easy and fully automatic with Secador® Auto-Desiccator Cabinets. The patented, electronic desiccant regeneration module recharges the desiccant every 20 minutes. Compared to other auto-desiccators that regenerate once every 4 to 6 hours, the Secador® Auto-Desiccator quickly attains a low %rH level on start-up and consistently maintains low %rH, especially with frequent entry to the desiccator interior.

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service