BAF420721009
SP Bel-Art® Secador® 2.0 Gas-Purge Desiccator Cabinet
volume 1.2 ft3
About This Item
Recommended Products
material
amber
polycarbonate
feature
Can be stacked three high
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F42072-1009
W × D
34.1 cm (13.4 in.) × 41.4 cm (16.3 in.)
volume
1.2 ft3
General description
- Durastar polycarbonate construction blocks 99% of UV light* and is resistant to staining, crazing and chemical attack
- Rear mounted gas ports are tapered to accept tubing with 1/4 to 3/4" I.D.
- Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
- Easy-to-view digital hygrometer is located in the door front ; side swinging door opens to the left
- Compact 34.1W x 41.4cmD footprint for space saving and can be stacked up to 3 units high
- Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves.
- Gas ports can be closed to use desiccator with standard desiccant cartridges
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)