B75204
2-Bromopentane
95%
Synonym(s):
(±)-2-Bromopentane, (±)-2-Pentyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH2CH2CHBrCH3
CAS Number:
Molecular Weight:
151.04
Beilstein:
1718829
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
liquid
refractive index
n20/D 1.441 (lit.)
bp
116-117 °C (lit.)
density
1.223 g/mL at 25 °C (lit.)
SMILES string
CCCC(C)Br
InChI
1S/C5H11Br/c1-3-4-5(2)6/h5H,3-4H2,1-2H3
InChI key
LGAJYTCRJPCZRJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
remainder 3-Bromopentane
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Flam. Liq. 2 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
68.0 °F - closed cup
Flash Point(C)
20 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Klaudia Chmelova et al.
Applied and environmental microbiology, 86(17) (2020-06-21)
Haloalkane dehalogenases can cleave a carbon-halogen bond in a broad range of halogenated aliphatic compounds. However, a highly conserved catalytic pentad composed of a nucleophile, a catalytic base, a catalytic acid, and two halide-stabilizing residues is required for their catalytic
Analysis of teratogenic effects of maternal treatment with 2-bromopropane in mice.
Hitoshi Ishikawa et al.
Journal of occupational health, 45(1), 63-65 (2003-11-08)
[Establishment of the maximum permissible concentration of 2-brompentane in the air of a work area].
M B Lis et al.
Gigiena truda i professional'nye zabolevaniia, (5)(5), 51-52 (1981-05-01)
S Sekiguchi et al.
Industrial health, 36(3), 297-299 (1998-08-14)
The influence of 2-bromopropane, an alternative to freon, on ovulation was examined using superovulation induced by pregnant mare's serum gonadotropin and human chorionic gonadotropin (PMSG-hCG) treatment. Three groups of mice were injected intraperitoneally 8 times with 2-bromopropane (2-BP) at 500
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service