B50805
1,4-Bis(5-phenyl-2-oxazolyl)benzene
suitable for scintillation, 98%
Synonym(s):
1,4-Bis(5-phenyl-2-oxazolyl)benzene, 2,2′-p-Phenylene-bis(5-phenyloxazole), Di(phenyl-5-oxazolyl-2)-1,4-benzene, POPOP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C24H16N2O2
CAS Number:
Molecular Weight:
364.40
Beilstein:
325741
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
Recommended Products
Assay
98%
mp
242-246 °C (lit.)
suitability
suitable for scintillation
SMILES string
c1ccc(cc1)-c2cnc(o2)-c3ccc(cc3)-c4ncc(o4)-c5ccccc5
InChI
1S/C24H16N2O2/c1-3-7-17(8-4-1)21-15-25-23(27-21)19-11-13-20(14-12-19)24-26-16-22(28-24)18-9-5-2-6-10-18/h1-16H
InChI key
MASVCBBIUQRUKL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
1,4-Bis(5-phenyl-2-oxazolyl)benzene (POPOP) is a scintillation luminophore. It is commonly used as a secondary wavelength shifter. The photochemical and photophysical studies of POPOP in different solvents have been reported.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Photophysical Parameters, Excitation Energy Transfer, and Photoreactivity of 1,4-Bis (5-phenyl-2-oxazolyl) benzene (POPOP) Laser Dye.
El-Daly SA, et al.
International Journal of Photoenergy, 2012 (2012)
Investigation of the physical and the optical properties of various base solvents for the liquid scintillator in a neutrino detector.
So SH, et al.
The Journal of the Korean Physical Society, 62(1), 26-33 (2013)
Kadir Aslan et al.
Analytical chemistry, 80(19), 7304-7312 (2008-09-04)
The use of zinc thin films deposited onto glass supports for surface plasmon coupled fluorescence (SPCF) over a broad 200 nm wavelength range is demonstrated. Fresnel calculations performed in the ultraviolet and visible spectral range are predicted to generate surface
N S Kudryasheva et al.
Luminescence : the journal of biological and chemical luminescence, 16(3), 243-246 (2001-06-16)
The involvement of upper electron-excited states in bacterial bioluminescence process was studied with excitation energy-accepting molecules. The fluorescent aromatic compounds, anthracene and 1.4-bis(5-phenyloxazol-2-yl)benzene, were chosen. Energies of their lowest excited singlet states are higher than the energy of the analogous
Xiaohui Hou et al.
Analytical biochemistry, 368(1), 100-110 (2007-06-15)
In this study, micron-sized poly(styrene-co-glycidyl methacrylate) (PSt-GMA) fluorescent microspheres of 5.1microm in diameter were synthesized via dispersion polymerization of styrene and glycidyl methacrylate in the presence of 1,4-bis(5-phenyloxazol-2-yl) benzene (POPOP), which provided surface functional groups for covalent immobilization of enzymes.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service