Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,4-Bis(5-phenyl-2-oxazolyl)benzene
Synonym(s):
1,4-Bis(5-phenyl-2-oxazolyl)benzene, 2,2′-
p
-Phenylene-bis(5-phenyloxazole), Di(phenyl-5-oxazolyl-2)-1,4-benzene, POPOP
Empirical Formula (Hill Notation):
C
24
H
16
N
2
O
2
CAS Number:
1806-34-4
Molecular Weight:
364.40
EC Number:
217-304-6
Products
Product Category
substrates (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
P3754
1,4-Bis(5-phenyl-2-oxazolyl)benzene,
BioReagent, suitable for scintillation
Expand
View Pricing