All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C10H8Br2N2
CAS Number:
Molecular Weight:
315.99
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
mp
152-156 °C (lit.)
SMILES string
BrCc1nc2ccccc2nc1CBr
InChI
1S/C10H8Br2N2/c11-5-9-10(6-12)14-8-4-2-1-3-7(8)13-9/h1-4H,5-6H2
InChI key
LHKFFORGJVELPC-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hyuk-Joon Jeon et al.
Biochemical and biophysical research communications, 489(2), 193-199 (2017-05-30)
Peroxiredoxins (Prxs) are highly conserved antioxidant enzymes and are implicated in multiple biological processes; however, their function in oocyte meiosis has not been studied. Here we show that inhibition of Prx I and II results in spindle defects, chromosome disorganization
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service