Skip to Content
Merck
All Photos(1)

Key Documents

A-LMPN100

Sigma-Aldrich

AnteoBind Micro

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$516.00

$516.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$516.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.22

$516.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

availability

not available in Japan

General description

AnteoBind Micro was developed for binding antibodies and other proteins to magnetic particles greater than 1 μm in size. It forms strong multi-valent interactions with almost any electron donating groups including carboxylic acids. Once proteins are added, AnteoBind activated particles rapidly form very strong but gentle binding interactions.
AnteoBind Micro has been used to stably activate many particles. Examples include: M-270 Dynabeads®, Bangs ProMag®, JSR MS300 Carboxyl and Estapor® M1-200/20.
AnteoBind Micro has many advantages over existing methods such as EDC/NHS chemistry or alternative ″pre-activated″ methods such as Epoxy or Tosyl. AnteoBind is water based and replaces the dry chemicals you would use with the EDC method. Using AnteoBind instead of NHS/EDC chemistry can save up to 3-4 hours of lab time. Experiments have shown increased stability and functionality for antibodies and a decreased background. AnteoBind improves reproducibility between experiments by providing uniformity between surfaces. For particles from 0.5 μm to 1 μm the recommended AnteoBind product is AnteoBind Sub-Micron.

Application

Activation and coupling of particles > 1 μm in diameter

Features and Benefits

  • Water based, activation and coupling reagent for particles > 1 μm in diameter
  • An alternative to traditional chemistries such as NHS/EDC methods with an easier to use AnteoBind protocol
  • Reduce reagent preparation time by 3-4 hours
  • Increase antibody functionality
  • Low background noise
  • High reproducibility between experiments

Legal Information

Anteobind is a trademark of AnteoTech Ltd
Dynabeads is a registered trademark of Invitrogen Dynal AS Corp
Estapor is a registered trademark of Merck KGaA, Darmstadt, Germany
ProMag is a registered trademark of Polysciences, Inc

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Key raw materials for lateral flow assays optimize IVD assay development for successful commercialization.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service