Skip to Content
Merck
All Photos(1)

Key Documents

ALD00586

Sigma-Aldrich

Pyridin-3-yl sulfurofluoridate

95%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$62.50

$62.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$62.50

About This Item

Empirical Formula (Hill Notation):
C5H4FNO3S
Molecular Weight:
177.15
UNSPSC Code:
12352200
NACRES:
NA.22

$62.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Pyridin-3-yl sulfurofluoridate can be used in the Pd-catalyzed Suzuki-Miyuara cross-coupling reaction. The fluorosulfate functionality was shown by Sharpless and coworkers to have distinct reactivity in comparison to other functional groups used in Suzuki-Miyuara reactions, such as bromides, chlorides and triflates. This inherent property provides a strong potential for selective Suzuki-Miyuara couplings for chemoselective reaction sequences.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Sharpless Lab pursues useful new reactivity and general methods for selectively controlling chemical reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service