Skip to Content
Merck
All Photos(2)

Key Documents

ALD00160

Sigma-Aldrich

2-Thiophenesulfonyl fluoride

95%

Synonym(s):

2-Thiophenesulfonyl fluoride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$138.00

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$138.00

About This Item

Empirical Formula (Hill Notation):
C4H3FO2S2
CAS Number:
Molecular Weight:
166.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

reaction suitability

reaction type: click chemistry

refractive index

n/D 1.509

density

1.437 at 25 °C

SMILES string

O=S(C1=CC=CS1)(F)=O

InChI

1S/C4H3FO2S2/c5-9(6,7)4-2-1-3-8-4/h1-3H

InChI key

NYJIUCJXMLNPHU-UHFFFAOYSA-N

Application

Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

223.0 °F

Flash Point(C)

106.11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service