Skip to Content
Merck
All Photos(1)

Key Documents

A59565

Sigma-Aldrich

6-Aminoindazole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N3
CAS Number:
Molecular Weight:
133.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

204-206 °C (dec.) (lit.)

SMILES string

Nc1ccc2cn[nH]c2c1

InChI

1S/C7H7N3/c8-6-2-1-5-4-9-10-7(5)3-6/h1-4H,8H2,(H,9,10)

InChI key

KEJFADGISRFLFO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Pinelli et al.
Arzneimittel-Forschung, 39(3), 361-365 (1989-03-01)
Administration of 6-aminoindazole (6-AIN) and 5-aminoindazole (5-AIN) to rats depressed gastric acid secretion, both basal and carbachol-stimulated. 6-AIN proved to be more active than 5-AIN in decreasing the stimulated secretory process. The antisecretory activity of 6-AIN appears to be partially
R Pabla et al.
Journal of molecular and cellular cardiology, 28(10), 2097-2110 (1996-10-01)
Nitric oxide (NO) is an endogenous protectant against reperfusion-induced ventricular fibrillation (VF) in the rat isolated heart. Here, the following were investigated: (1) the tissue source of cardioprotective NO using a novel inhibitor (7-nitro indazole; 7-NI) of the neuronal form

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service