Skip to Content
Merck
All Photos(1)

Key Documents

A45009

Sigma-Aldrich

Ethyl 3-aminopyrazole-4-carboxylate

98%

Synonym(s):

3-Amino-4-carbethoxypyrazole

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$44.00

$44.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$44.00

About This Item

Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$44.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

powder

mp

105-107 °C (lit.)

SMILES string

CCOC(=O)c1c[nH]nc1N

InChI

1S/C6H9N3O2/c1-2-11-6(10)4-3-8-9-5(4)7/h3H,2H2,1H3,(H3,7,8,9)

InChI key

YPXGHKWOJXQLQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gemma N Ferguson et al.
Journal of medicinal chemistry, 51(19), 6165-6172 (2008-09-06)
A pharmacophore-based screen identified 32 compounds including ethyl 5-amino-3-(4- tert-butylphenyl)-4-oxo-3,4-dihydrothieno[3,4- d]pyridazine-1-carboxylate ( 8) as a new allosteric modulator of the adenosine A1 receptor (A1AR). On the basis of this lead, various derivatives were prepared and evaluated for activity at the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service