Skip to Content
Merck
All Photos(1)

Key Documents

705969

Sigma-Aldrich

2-Methyloxazole-4-carboxylic acid

97%

Synonym(s):

2-Methyl-1,3-oxazole-4-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$70.00

$70.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$70.00

About This Item

Empirical Formula (Hill Notation):
C5H5NO3
CAS Number:
Molecular Weight:
127.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$70.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

97%

form

solid

mp

182-187 °C

functional group

carboxylic acid

SMILES string

Cc1nc(co1)C(O)=O

InChI

1S/C5H5NO3/c1-3-6-4(2-9-3)5(7)8/h2H,1H3,(H,7,8)

InChI key

IARMCEYEYXXEOS-UHFFFAOYSA-N

Application

Reactant for:
  • Solid-phase preparation of (heteroaryl)carbonyl-substituted dipeptides as human protease activated receptor 2 affectors
  • Pd-catalyzed decarboxylative C-H cross-coupling
  • Stereoselective preparation of 3-amino-N-(phenylsulfonyl)alanines as inhibitors of ανβ3 integrin

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service